Learn More
Lung Carcinoma, Mouse, Clone: CHALU2, Abnova™
Mouse monoclonal antibody raised against Lung Carcinoma.
Manufacturer:  Abnova Corporation MAB6361
Catalog No. 89-136-191
Add to cart
Description
Description
Specifications
Specifications
Lung Carcinoma
CHALU2
Mouse monoclonal antibody raised against Lung Carcinoma.
Lyophilized from 20mM PBS, pH 7.0
Mouse
IgG1, kappa
RUO
Primary
Human
ELISA, Immunofluorescence
Unconjugated
The optimal working dilution should be determined by the end user.
Lyophilized
Native from human lung carcinoma cells.
500 ug
Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Monoclonal