Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LXR alpha/NR1H3 Antibody (OTI1A5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP246220
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
LXR alpha/NR1H3 Monoclonal specifically detects LXR alpha/NR1H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LXR alpha/NR1H3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin | |
alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
Mouse | |
50.2 kDa | |
0.1 mL | |
Diabetes Research, Lipid and Metabolism | |
10062 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
OTI1A5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
NR1H3 | |
Human recombinant protein fragment corresponding to amino acids 69-329 of human NR1H3 (NP_005684) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction