Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MB, Mouse, Clone: 7C3, Abnova™

Mouse monoclonal antibody raised against native MB.

Manufacturer:  Abnova Corporation MAB0565

Catalog No. 89-107-982

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a haemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq

Specifications

Specifications

MB
7C3
Mouse monoclonal antibody raised against native MB.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
MB
Native purified human MB.
200 ug
Store at 4°C.
Monoclonal
Human
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
MB
MGC13548/PVALB
Mouse
IgG1
RUO
Primary
4151
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit