Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFG-E8 Antibody (EDM45), Novus Biologicals™
Mouse Monoclonal Antibody
$265.00 - $506.50
Specifications
Antigen | MFG-E8 |
---|---|
Clone | EDM45 |
Concentration | 0.2mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24441200
|
Novus Biologicals
NBP2444120.02MG |
0.02 mg |
Each for $265.00
|
|
|||||
NBP24441201
|
Novus Biologicals
NBP2444120.1MG |
0.1 mg |
Each for $506.50
|
|
|||||
NBP24441202
|
Novus Biologicals
NBP2444120.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
MFG-E8 | |
0.2mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, medin, MFG1, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
MFGE8 | |
IgG1 κ | |
Protein A or G purified | |
Recognizes a protein of 40-45kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development. |
EDM45 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q08431 | |
4240 | |
Delipidated human milk fat globule membrane preparation | |
Primary | |
Store at 4C. | |
45 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title