Learn More
Mouse anti-MHC Class I, Clone: 3D12HLA-E, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1431240.025MG
 View more versions of this product
Catalog No. NBP14312403
Add to cart
Description
Description
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.
Specifications
Specifications
HLA-E
3D12HLA-E
Unconjugated
PBS, pH 7.2 with 0.09% Sodium Azide
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Mouse
IgG1 Kappa
0.025 mg
Adaptive Immunity, Immunology, Major Histocompatibility Complexes
Primary
3133
Flow Cytometry, Immunoprecipitation, Western Blot
1.0 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Affinity Purified
HLA-E
The immunogen for this antibody was HLAE.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human