Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIZF, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant MIZF.

Manufacturer:  Abnova Corporation H00025988A01

Catalog No. 89-106-674


Add to cart

Description

Description

MIZF interacts with methyl-CpG-binding protein-2 (MBD2; MIM 603547), a component of the MeCP1 histone deacetylase (HDAC) complex, and plays a role in DNA methylation and transcription repression.[supplied by OMIM

Sequence: MPPPGKVPRKENLWLQCEWGSCSFVCSTMEKFFEHVTQHLQQHLHGSGEEEEEEEEDDPLEEEFSCLWQECGFCSLDSSADLIRHVYFHCYHTKLKQWGLQALQSQADLG
Specifications

Specifications

MIZF
Unconjugated
50% glycerol
NM_015517
DKFZp434F162/HiNF-P/ZNF743
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant MIZF.
MIZF
Antisera
MIZF
MIZF (NP_056332, 1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag.
RUO
Primary
25988
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only