Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-12 Antibody (603E6.22), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals DDX0280HRPO100
Description
MMP-12 Monoclonal specifically detects MMP-12 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
MMP-12 | |
Monoclonal | |
0.5mg/mL | |
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen | |
EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP-12 | |
Mouse | |
Ion exchange chromatography | |
RUO | |
4321 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
603E6.22 | |
HRP | |
PBS and 50% Glycerol with No Preservative | |
MMP12 | |
MMP-12/HME was used as immunogen. | |
0.1 mg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction