Learn More
CD45.1 Mouse anti-Mouse, Brilliant Violet 786, Clone: A20, BD Optibuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 740889
Catalog No. BDB740889
Add to cart
Description
Description
The A20 monoclonal antibody specifically binds to CD45 (Leukocyte Common Antigen) on all leukocytes of mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, DA). This alloantigen was originally named Ly-5.2, but was later changed to Ly-5.1 to conform with the convention that the .2 alloantigen designates the C57BL/6 strain. mAb A20 has been reported not to react with leukocytes of most other mouse strains which express the CD45.2 alloantigen. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family; its intracellular (COOH-terminal) region contains two PTP catalytic domains while the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), and differing levels of glycosylation. CD45 isoforms in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The A20 antibody has been reported to inhibit some responses of B cells (from mice expressing the CD45.1 alloantigen) to antigens and LPS.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
Specifications
CD45.1
A20
Brilliant Violet 786
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Ly-5; Lyt4; CD45R; LCA; Ptprc; Protein tyrosine phosphatase receptor type C
Affinity Purified
SJL mouse thymocytes and splenocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine