Learn More
MRC2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MRC2.
Supplier: Abnova Corporation H00009902A01
Description
MRC2 is a recycling endocytic receptor that functions in cell motility and remodeling of the extracellular matrix by promoting cell migration and uptake of collagens for intracellular degradation (Wienke et al., 2007 [PubMed 17974964]).[supplied by OMIM
Sequence: ASLGMYECDREALNLRWHCRTLGDQLSLLLGARTSNISKPGTLERGDQTRSGQWRIYGSEEDLCALPYHEVYTIQGNSHGKPCTIPFKYDNQWFHGCTSTGREDGHLWCASpecifications
MRC2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MRC2. | |
MRC2 | |
CD280/CLEC13E/ENDO180/FLJ35911/KIAA0709/UPARAP | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006039 | |
MRC2 | |
MRC2 (NP_006030, 105 a.a. ∽ 214 a.a) partial recombinant protein with GST tag. | |
RUO | |
9902 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only