Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-MRP1, Clone: MRPm6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1423401.0ML

Catalog No. NBP14234010


Add to cart

Description

Description

MRP1 Monoclonal antibody specifically detects MRP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

MRP1
MRPm6
Western Blot 1:20-1:50, Flow Cytometry 1:20-1:50, Immunohistochemistry 1:20-50, Immunohistochemistry-Paraffin 1:20-50, Immunohistochemistry-Frozen 1:20-50
Supernatant
ABCC1
The antibody reacts with the MRP1 antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
MRPm6 reacts with an internal epitope of MRP, a 180-195 kD transmembrane transporter protein overexpressed in various human non-P-glycoprotein MDR tumor cell lines. MRPm6 was raised against a bacterial fusion protein of MRP, containing a segment of 170 amino acids in the carboxy terminal end and part of the carboxy proximal nucleotide binding domain of the protein. MRPm6 does not cross react with the human MDR1 and MDR3 gene products (Flens et al. 1994)
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
No buffer with 0.1% Sodium Azide
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein
Mouse
IgG1
1 ml
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction
Primary
4363
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only