Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MTA1, Mouse, Clone: 1F12, Abnova™

Mouse monoclonal antibody raised against a partial recombinant MTA1.

Manufacturer:  Abnova Corporation H00009112M13A

Catalog No. 89-106-011


Add to cart

Description

Description

This gene encodes a protein that was identified in a screen for genes expressed in metastatic cells, specifically, mammary adenocarcinoma cell lines. Expression of this gene has been correlated with the metastatic potential of at least two types of carcinomas although it is also expressed in many normal tissues. The role it plays in metastasis is unclear. It was initially thought to be the 70kDa component of a nucleosome remodeling deacetylase complex, NuRD, but it is more likely that this component is a different but very similar protein. These two proteins are so closely related, though, that they share the same types of domains. These domains include two DNA binding domains, a dimerization domain, and a domain commonly found in proteins that methylate DNA. The profile and activity of this gene product suggest that it is involved in regulating transcription and that this may be accomplished by chromatin remodeling. [provided by RefSeq

Sequence: MPSRGLANHGQTRHMGPSRNLLLNGKSYPTKVRLIRGGSLPPVKRRRMNWIDAPDDVFYMATEETRKIRKLLSSSETKRAARRPYKPIALRQSQALPPRP
Specifications

Specifications

MTA1
Monoclonal
Unconjugated
ascites with no preservative
NM_004689
MTA1
MTA1 (NP_004680,601 a.a. ∽ 700 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
200 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
9112
ELISA, Western Blot
1F12
Mouse monoclonal antibody raised against a partial recombinant MTA1.
MTA1
Ascites
Mouse
IgG1 Kappa
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit