Learn More
MUC1, Mouse, Clone: M3A106, Abnova™
Mouse monoclonal antibody raised against native MUC1.
Manufacturer:  Abnova Corporation MAB0553
Catalog No. 89-107-980
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene is a member of the mucin family and encodes a membrane bound, glycosylated phosphoprotein. The protein is anchored to the apical surface of many epithelia by a transmembrane domain, with the degree of glycosylation varying with cell type. It also includes a 20 aa variable number tandem repeat (VNTR) domain, with the number of repeats varying from 20 to 120 in different individuals. The protein serves a protective function by binding to pathogens and also functions in a cell signaling capacity. Overexpression, aberrant intracellular localization, and changes in glycosylation of this protein have been associated with carcinomas. Multiple alternatively spliced transcript variants that encode different isoforms of this gene have been reported, but the full-length nature of only some has been determined. [provided by RefSeq
Specifications
Specifications
MUC1
M3A106
Mouse monoclonal antibody raised against native MUC1.
In PBS, pH 7.4 (0.09% sodium azide)
Liquid
MUC1
Native purified human MUC1.
200 ug
Store at 4°C.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Frozen)
Unconjugated
The optimal working dilution should be determined by the end user.
MUC1
CD227/EMA/H23AG/MAM6/PEM/PEMT/PUM
Mouse
IgG
RUO
Primary
4582