Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MUC5B, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MUC5B.
Supplier: Abnova Corporation H00727897A01
Description
Mucins are high molecular mass, highly glycosylated macromolecules that are the major components of mucus secretions. MUC5B is a salivary mucin that is thought to contribute to the lubricating and viscoelastic properties of whole saliva. It is composed of 14.9% protein, 78.1% carbohydrate, and 7% sulfate (Troxler et al., 1995 [PubMed 8554565]).[supplied by OMIM
Sequence: CEEDSCQVRINTTILWHQGCETEVNITFCEGSCPGASKYSAEAQAMQHQCTCCQERRVHEETVPLHCPNGSAILHTYTHVDECGCTPFCVPAPMAPPHTRGFPAQEATAVSpecifications
MUC5B | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant MUC5B. | |
MUC5B | |
MG1/MUC5/MUC9 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
XM_039877 | |
MUC5B | |
MUC5B (XP_039877.9, 4186 a.a. ∽ 4295 a.a) partial recombinant protein with GST tag. | |
RUO | |
727897 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction