Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Mucin 5AC, Clone: 45M1, Novus Biologicals™

Mouse Monoclonal Antibody

$421.50

Specifications

Antigen Mucin 5AC
Clone 45M1
Concentration 0.2mg/mL
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
Gene Accession No. P98088
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP21519601
View Documents
Novus Biologicals
NBP2151960.1MG
0.1 mg
Each for $421.50
Add to cart
 
NBP21519602
View Documents
Novus Biologicals
NBP2151960.2MG
0.2 mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

MUC5AC Monoclonal antibody specifically detects MUC5AC in Human, Mouse, Rat, Porcine, Chicken, Feline, Primate, Rabbit, Bovine (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications

Specifications

Mucin 5AC
0.2mg/mL
P98088
PBS with 0.05% BSA. with 0.05% Sodium Azide
Monoclonal
Store at 4C.
IgG1 Kappa
Purified
RUO
45M1
M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b) patient
4586
This MAb recognizes the peptide core of gastric mucin M1 (>1,000kDa) (recently identified as Mucin 5AC). Its epitope is destroyed by beta-mercaptoethanol and proteases but not by periodate treatment. Antibody to gastric mucin M1 reacts with the gastric epithelium of normal human gastrointestinal tract as well as with the precancerous and cancerous colon but not with normal adult colon. It also reacts with fetal colonic mucosa. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.
Primary
Mouse
Unconjugated
Protein G purified
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit