Learn More
Mouse anti-Myc Epitope Tag, Clone: 2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243627
Catalog No. NBP243627
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Myc Epitope Tag Monoclonal antibody specifically detects Myc Epitope Tag in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
Myc Epitope Tag
2
Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Immunoprecipitation 1:100-1:500
Purified
The exact sequence is proprietary.
Protein G purified
RUO
Primary
Human
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
PBS with No Preservative
Mouse
IgG1
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal