Learn More
Mouse anti-Napsin-A, Clone: TMU-Ad02, Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11068133AF488
Catalog No. NB11068133X
Add to cart
Description
Description
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Napsin A
TMU-Ad02
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
O96009
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
Mouse
IgG1
0.1 ml
Cancer, Cell Biology, Cellular Markers
Primary
9476
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
NAPSA
Synthetic peptide corresponding to the N-terminus of human NAPSA [UniProt# O96009]
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Protein A purified