Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT6, Mouse, Clone: AT2F4, Abnova™
Mouse monoclonal antibody raised against full-length recombinant NAT6.
Supplier: Abnova Corporation MAB7884
Specifications
NAT6 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
NAT6 | |
NAT6 | |
Recombinant protein corresponding to amino acids 1-308 of human NAT6. | |
RUO | |
24142 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
AT2F4 | |
Mouse monoclonal antibody raised against full length recombinant NAT6. | |
In PBS, pH 7.4 (0.09% sodium azide) | |
FUS-2/FUS2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction