Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-NCK1, Clone: 6B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242644

Catalog No. NBP242644


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

NCK1 Monoclonal antibody specifically detects NCK1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

NCK1
6B3
Unconjugated
PBS (pH 7.4) and 10% Glycerol with 0.02% Sodium Azide
Purified
NCK1
Recombinant human NCK1 (1-377aa) purified from E. coli
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ELISA
1 mg/mL
Western Blot 1:2000, ELISA
NP_006144
cytoplasmic protein NCK1, MGC12668, NCK adaptor protein 1melanoma NCK protein, nck-1, NCKalpha, NCKNCK tyrosine kinase, non-catalytic region of tyrosine kinase, SH2/SH3 adaptor protein NCK-alpha
Mouse
IgG3 Kappa
100 ul
Cancer, Cell Cycle and Replication
Primary
4690
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only