Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA6, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant NCOA6.

Manufacturer:  Abnova Corporation H00023054A01

Catalog No. 89-106-550


Add to cart

Description

Description

The protein encoded by this gene is a transcriptional coactivator that can interact with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has been shown to be involved in the hormone-dependent coactivation of several receptors, including prostanoid, retinoid, vitamin D3, thyroid hormone, and steroid receptors. The encoded protein may also act as a general coactivator since it has been shown to interact with some basal transcription factors, histone acetyltransferases, and methyltransferases. [provided by RefSeq

Sequence: VGSHPELLPSIAPSQNLVSKETSTTALQASVARPELEVNAAIVSGQSSEPKEIVEKSKIPGRRNSRTEEPTVASESVENGHRKRSSRPASASSSTKDITSAVQSKRRKSK
Specifications

Specifications

NCOA6
Unconjugated
50% glycerol
NM_014071
AIB3/ANTP/ASC2/HOX1.1/HOXA7/KIAA0181/NRC/PRIP/RAP250/TRBP
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Mouse polyclonal antibody raised against a partial recombinant NCOA6.
NCOA6
Antisera
NCOA6
NCOA6 (NP_054790, 1954 a.a. ∽ 2063 a.a) partial recombinant protein with GST tag.
RUO
Primary
23054
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only