Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-NCOR2, Polyclonal, Novus Biologicals™

Mouse Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243699

Catalog No. NBP243699


Add to cart

Description

Description

Specifically detects NCOR2 in Human samples, and it is validated for Immunoprecipitation, Western Blotting

NCOR2 Polyclonal antibody specifically detects NCOR2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

NCOR2
Unconjugated
PBS (pH 7.4) with No Preservative
CTG repeat protein 26, CTG26nuclear receptor co-repressor 2, FLJ25011, N-CoR2, nuclear receptor corepressor 2, retinoic-acid-receptor-associated corepressor, silencing mediator for retinoid and thyroid hormone receptors, SMAP270, SMRTE, SMRTE-tau, SMRTT3 receptor-associating factor, TNRC14, TRAC, TRAC1
Mouse
IgG
50 ul
Cancer, Chromatin Research, Transcription Factors and Regulators
Primary
9612
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Purified
NCOR2
A synthetic peptide from the human SMRT protein corresponding to amino acids 80-92 (QELHLRPESHSYL).
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only