Learn More
NFKBIL2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant NFKBIL2.
Supplier: Abnova Corporation H00004796A01
Description
The protein encoded by this gene is thought to be a negative regulator of NF-kappa-B mediated transcription. The encoded protein may bind NF-kappa-B complexes and trap them in the cytoplasm, preventing them from entering the nucleus and interacting with the DNA. Phosphorylation of this protein targets it for degradation by the ubiquitination pathway, which frees the NF-kappa-B complexes to enter the nucleus. [provided by RefSeq
Sequence: MRTRLYLNLGLTFESLQQTALCNDYFRKSIFLAEQNHLYEDLFRARYNLGTIHWRAGQHSQAMRCLEGARECAHTMRKRFMESECCVVIAQVLQDLGSpecifications
NFKBIL2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant NFKBIL2. | |
NFKBIL2 | |
FLJ40087/IKBR | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_013432 | |
NFKBIL2 | |
NFKBIL2 (NP_038460, 1 a.a. ∽ 97 a.a) partial recombinant protein with GST tag. | |
RUO | |
4796 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only