Learn More
Mouse anti-NGFI-B alpha/Nur77/NR4A1, Clone: 12.14, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1432690.025MG
Catalog No. NBP14326903
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
NGFI-B alpha/Nur77/NR4A1
12.14
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Early response protein NAK1, GFRP1ST-59, growth factor-inducible nuclear protein N10, HMRNP10, hormone receptor, MGC9485, N10, NAK1, NAK-1, NGFIB, Nuclear hormone receptor NUR/77, nuclear receptor subfamily 4 group A member 1, nuclear receptor subfamily 4, group A, member 1, NUR77, Orphan nuclear receptor HMR, Orphan nuclear receptor TR3, steroid receptor TR3, Testicular receptor 3, TR3, TR3 orphan receptor
Mouse
IgG1
0.025 mg
Apoptosis
Primary
3164
Western Blot, Flow Cytometry
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry
Affinity Purified
NR4A1
The immunogen for this antibody was Nur77.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse