Learn More
Mouse anti-NHE8, Clone: 7A11, Alexa Fluor 647, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11062091AF647
Catalog No. NB11062091Y
Add to cart
Description
Description
NHE8 Monoclonal specifically detects NHE8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
NHE8
7A11
Western Blot, Immunocytochemistry/Immunofluorescence
Q9Y2E8
DKFZp686C03237, KIAA0939DKFZp686C03237, MGC138418, Na(+)/H(+) exchanger 8, NHE-8, NHE8FLJ42500, sodium/hydrogen exchanger 8, solute carrier family 9 (sodium/hydrogen exchanger), solute carrier family 9 (sodium/hydrogen exchanger), isoform 8, solute carrier family 9 (sodium/hydrogen exchanger), member 8, Solute carrier family 9 member 8
Mouse
IgG
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This is specific for the 85 kDa mature glycosylated form of the protein. It does not recognize the 55 kDa immature form.
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SLC9A8
A fusion protein containing the C-terminal 89 residues of human NHE8. [Swiss-Prot# Q9Y2E8]
80 kDa
0.1 ml
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism
Primary
23315
Human, Mouse, Rat