Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Nuclear Membrane Marker, Clone: NM97, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Nuclear Membrane Marker |
---|---|
Clone | NM97 |
Concentration | 1.0 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml |
Applications | Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23469602
|
Novus Biologicals
NBP2346960.2 MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Nuclear Membrane Marker Monoclonal antibody specifically detects Nuclear Membrane Marker in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Nuclear Membrane Marker | |
1.0 mg/mL | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Human | |
Nuclei of myeloid leukemia biopsy cells | |
Primary | |
Protein A purified | |
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the nuclear membrane expressed in human cells. It can be used to stain the nuclear membrane in cell or tissue preparations and can be used as a marker of the nuclear membrane in subcellular fractions. It produces a ring pattern around the nucleus of cells of normal and malignant cells and may be used to stain the nuclear membrane of cells in fixed or frozen tissue sections.,The nuclear envelope (also known as the perinuclear envelope, nuclear membrane, nucleolemma or karyotheca) is the double membrane of the nucleus that encloses genetic material in eukaryotic cells. It separates the contents of the nucleus (DNA in particular) from the cytosol (cytoplasm). Numerous nuclear pores are present on the nucl |
NM97 | |
Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS - BSA free with No Preservative | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title