Learn More
NUF2 (CDCA1), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant CDCA1.
Supplier: Abnova Corporation H00083540A01
Description
This gene encodes a protein that is highly similar to yeast Nuf2, a component of a conserved protein complex associated with the centromere. Yeast Nuf2 disappears from the centromere during meiotic prophase when centromeres lose their connection to the spindle pole body, and plays a regulatory role in chromosome segregation. The encoded protein is found to be associated with centromeres of mitotic HeLa cells, which suggests that this protein is a functional homolog of yeast Nuf2. Alternatively spliced transcript variants that encode the same protein have been described. [provided by RefSeq
Sequence: METLSFPRYNVAEIVIHIRNKILTGADGKNLTKNDLYPNPKPEVLHMIYMRALQIVYGIRLEHFYMMPVNSEVMYPHLMEGFLPFSNLVTHLDSFLPICRVNDFETADILCPKAKRTSRFLSGIINFIHFREACRETYMEFLWQYKSSADKMQQLNAAHQEALMKLERLDSVPVEEQEEFKQLSDGIQELQQSLNQDFHQKTIVLQEGNSQKKSNISEKTKRLNELKLSVVSLKEIQESLKTKIVDSPEKLKNYKEKMKDTVQKLKNARQEVVEKYEIYGDSVDCLPSCQLEVQLYQKKIQDLSDNREKLASILKESLNLEDQIESDESELKKLKTEENSFKRLMIVKKEKLATAQFKINKKHEDVKQYKRTVIEDCNKVQEKRGAVYERVTTINQEIQKIKLGIQQLKDAAEREKLKSQEIFLNLKTALEKYHDGIEKAAEDSYAKIDEKTAELKRKMFKMSTSpecifications
NUF2 (CDCA1) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant CDCA1. | |
NUF2 | |
CDCA1/NUF2R | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC008489 | |
NUF2 | |
CDCA1 (AAH08489, 1 a.a. ∽ 464 a.a) full-length recombinant protein with GST tag. | |
RUO | |
83540 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only