Learn More
Mouse anti-OATP1B3/SLCO1B3/OATP8, Alexa Fluor 700, Clone: 1F6.1B1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236493AF700
Catalog No. NBP236493Z
Add to cart
Description
Description
OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
OATP1B3/SLCO1B3/OATP8
1F6.1B1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Q9NPD5
Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3
Mouse
IgG3 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Purified
SLCO1B3
Two synthetic peptides made to the human SLCO1B3 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Protein G purified
RUO
Primary
28234