Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-OATP1B3/SLCO1B3/OATP8, APC, Clone: 1E6.2F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236492APC

Catalog No. NBP236492J


Add to cart

Description

Description

OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

OATP1B3/SLCO1B3/OATP8
1E6.2F9
Flow Cytometry
Q9NPD5
Liver-specific organic anion transporter 2, liver-specific organic anion transporter 3TM13, LST2, LST-2, LST3, OATP1B3LST-3TM13, OATP8, OATP-8, Organic anion transporter 8, organic anion transporter LST-3c, Organic anion-transporting polypeptide 8, SLC21A8, solute carrier family 21 (organic anion transporter), member 8, Solute carrier family 21 member 8, solute carrier organic anion transporter family member 1B3, solute carrier organic anion transporter family, member 1B3
Mouse
IgG3 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
Purified
SLCO1B3
Two synthetic peptides made to the human OATP8 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Protein G purified
RUO
Primary
28234
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit