Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-OATP1B3/SLCO1B3/OATP8, Clone: 1E6.2F9, Novus Biologicals™

Mouse Monoclonal Antibody

$119.60 - $318.00

Specifications

Antigen OATP1B3/SLCO1B3/OATP8
Clone 1E6.2F9
Concentration 1.0 mg/mL
Immunogen Two synthetic peptides made to the human OATP8 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
Gene Accession No. Q9NPD5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP236492SS
View Documents
Novus Biologicals
NBP236492SS
0.025 mg
Each for $119.60
Add to cart
 
NBP236492
View Documents
Novus Biologicals
NBP236492
0.1 mg
Each for $318.00
Add to cart
 
Description

Description

OATP1B3/SLCO1B3/OATP8 Monoclonal antibody specifically detects OATP1B3/SLCO1B3/OATP8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

OATP1B3/SLCO1B3/OATP8
1.0 mg/mL
Q9NPD5
PBS with 0.05% Sodium Azide
Primary
Mouse
Unconjugated
Protein G purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
1E6.2F9
Two synthetic peptides made to the human OATP8 protein sequence (between residues 648-700). [Swiss-Prot Q9NPD5]
28234
Monoclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG3 Kappa
Purified
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit