Learn More
Oligodendrocytes/Myelin (CNS), Mouse, Clone: HPC-7, Abnova™
Mouse monoclonal antibody raised against native Oligodendrocytes/Myelin (CNS).
Manufacturer:  Abnova Corporation MAB4861
Catalog No. 89-135-836
Description
Description
Specifications
Specifications
Oligodendrocytes/Myelin (CNS)
HPC-7
Mouse monoclonal antibody raised against native Oligodendrocytes/Myelin (CNS).
In 10mM PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Mouse
IgG1
RUO
Primary
Stains Oligodendrocytes and myelin in the CNS.
Immunohistochemistry
Unconjugated
Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user.
Liquid
Native purified from adult rat hippocampal membranes.
125 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Rat