Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPRK1, Mouse, Clone: 5E3, Abnova™
Mouse monoclonal antibody raised against OPRK1.
Supplier: Abnova Corporation MAB1740
Specifications
OPRK1 | |
Monoclonal | |
Unconjugated | |
Western Blot (5 ug/mL) Immunoprecipitation (1 ug/mL) Flow Cyt (1 ug/mL) Immunoprecipitation (5 ug/mL) The optimal working dilution should be determined by the end user. | |
OPRK1 | |
OPRK1 | |
hKOR expressing cell line. | |
RUO | |
4986 | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot | |
5E3 | |
Mouse monoclonal antibody raised against OPRK1. | |
In PBS (0.1% proclin, 2.0% Block Ace) | |
KOR/OPRK | |
Mouse | |
25 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction