Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAN2 (USP52), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant USP52.

Manufacturer:  Abnova Corporation H00009924A01

Catalog No. 89-106-172


Add to cart

Description

Description

Sequence: TVYLFHMPRKRMISLRFLAWYFLDLKIQGETHDSIEDARTALQLYRKYLELSKNGTEPESFHKVLKGLYEKGRKMDWKVPEPEGQTSPKNAAVFSSVLAL
Specifications

Specifications

PAN2 (USP52)
Unconjugated
50% glycerol
NM_014871
FLJ39360/KIAA0710/USP52/hPAN2
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant USP52.
PAN2
Antisera
PAN2
USP52 (NP_055686, 1099 a.a. ∽ 1198 a.a) partial recombinant protein with GST tag.
RUO
Primary
9924
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only