Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Pax3, Clone: 7D8G7, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237386

Catalog No. NBP237386

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Pax3
7D8G7
Unconjugated
PBS with 0.05% Sodium Azide
CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Mouse
IgG1
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot, ELISA
1 mg/mL
Western Blot 1:500 - 1:2000, ELISA 1:10000
Affinity Purified
PAX3
Purified recombinant fragment of human PAX3 (AA: 142-203) expressed in E. Coli.
53 kDa
0.1 ml
Apoptosis, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary
5077
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only