Learn More
PDIA6, Mouse, Clone: 3B4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant PDIA6.
Supplier: Abnova Corporation H00010130M04A
Description
Protein disulfide isomerases (EC 5.3.4.1), such as PDIA6, are endoplasmic reticulum (ER) resident proteins that catalyze formation, reduction, and isomerization of disulfide bonds in proteins and are thought to play a role in folding of disulfide-bonded proteins (Hayano and Kikuchi, 1995 [PubMed 7590364]).[supplied by OMIM
Sequence: MALLVLGLVSCTFFLAVNGLYSSSDDVIELTPSNFNREVIQSDSLWLVEFYAPWCGHCQRLTPEWKKAATALKDVVKVGAVDADKHHSLGGQYGVQGFPTIKIFGSNKNRPEDYQGGRTGEAIVDAALSALRQLVKDRLGGRSGGYSSGKQGRSDSSSKKDVIELTDDSFDRNVLDSEDVWMVEFYAPWCGHCKNLEPEWAAAASEVKEQTKGKVKLAAVDATVNQVLASRYGIRGFPTIKIFQKGESPVDYDGGRTRSDIVSRALDLFSDNAPPPELLEIINEDIAKRTCEEHQLCVVAVLPHILDTGAAGRNSYLEVLLKLADKYKKKMWGWLWTEAGAQSELETALGIGGFGYPAMAAINARKMKFALLKGSFSEQGINEFLRELSFGRGSTAPVGGGAFPTIVEREPWDGRDGELPVEGDIDLSDVELDDLGKDELSpecifications
PDIA6 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC001312 | |
PDIA6 | |
PDIA6 (AAH01312,1 a.a. ∽ 440 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
10130 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3B4 | |
Mouse monoclonal antibody raised against a full-length recombinant PDIA6. | |
PDIA6 | |
ERP5/P5/TXNDC7 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.