Learn More
Penicillin, Mouse, Clone: Pen-9, Abnova™
Mouse monoclonal antibody raised against Penicillin.
Manufacturer:  Abnova Corporation MAB2462
Catalog No. 89-135-199
Add to cart
Description
Description
Specifications
Specifications
Penicillin
Pen-9
Mouse monoclonal antibody raised against Penicillin.
In PBS (0.09% sodium azide)
Mouse
IgG1
RUO
Primary
The thiazolidine ring of penicillins.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Benzylpenicillin conjugated with bovine gamma-globulin.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bovine