Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perlecan Mouse anti-Human, Alexa Fluor 647, Clone: 5D7-2E4, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565782

Catalog No. BDB565782


Add to cart

Description

Description

The 5D7-2E4 monoclonal antibody specifically recognizes the protein core of Perlecan (PRCAN), which has a calculated molecular mass of ~460 kDa. Smaller molecular mass species of Perlecan have also been described. Perlecan is also known as Heparan sulfate proteoglycan 2 (HSPG2), or Basement membrane-specific heparan sulfate proteoglycan core protein (HSPG). Perlecan is a large multidomain proteoglycan that is produced by epithelial and endothelial cells. It is a major component of the vascular extracellular matrix and helps sustain endothelial barrier function. Perlecan inhibits smooth muscle cell proliferation and serves as an attachment substrate for cells. It plays essential roles in normal cartilage, heart and vascular development, as well as, in the response to vascular injury.

Specifications

Specifications

Perlecan
5D7-2E4
Alexa Fluor 647
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
PRCAN; HSPG2; HSPG; Endorepellin; LG3; PLC; SJA; SJS; SJS1
Affinity Purified
Purified full-length Human Perlecan
Affinity Chromatography
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit