Learn More
Mouse anti-PEX19, Clone: 533, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243757
Catalog No. NBP243757
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
PEX19 Monoclonal antibody specifically detects PEX19 in Human, Rat samples. It is validated for Western Blot.
Specifications
Specifications
PEX19
533
Western Blot 1:500-1:3000
Purified
PEX19
Recombinant protein encompassing a sequence within the center region of human PEX19. The exact sequence is proprietary.
33 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human, Rat
Western Blot
Unconjugated
PBS with No Preservative
D1S2223Ehousekeeping gene, 33kD, HK3333 kDa housekeeping protein, Peroxin-19, peroxisomal biogenesis factor 19, Peroxisomal farnesylated proteinFLJ55296, PMP1, PMPI, PXFperoxin-19, PXMP1
Mouse
IgG2b
Protein G purified
RUO
Primary
5824