Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenytoin, Mouse, Clone: 303, Abnova™

Mouse monoclonal antibody raised against Phenytoin.

Manufacturer:  Abnova Corporation MAB4282

Catalog No. 89-135-685

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Phenytoin
303
Mouse monoclonal antibody raised against Phenytoin.
In PBS, pH 7.4
Mouse
IgG1, kappa
RUO
Primary
Phenytoin
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Phenytoin conjugated with KLH.
1 mg
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit