Learn More
Mouse anti-Phospho-Tyrosine, Clone: 21, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242911
Catalog No. NBP242911
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Phospho-Tyrosine
21
Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:1000
Purified
Mouse
IgG2b
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS with No Preservative
2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, Tyrosine
The exact sequence is proprietary.
Protein G purified
RUO
Primary
Human