Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PNPT1 Antibody (3H5), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243712
Description
Ensure accurate, reproducible results in Immunocytochemistry/Immunofluorescence, Immunoprecipitation
PNPT1 Monoclonal specifically detects PNPT1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PNPT1 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
PNPT1 | |
The immunogen used to generate this antibody corresponds to human PNPase. | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunofluorescence | |
3H5 | |
Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
DKFZp762K1914, EC 2.7.7, EC 2.7.7.8, mitochondrial, old-35, OLD35, PNPase, PNPase 1, PNPase old-35, Polynucleotide phosphorylase 1, Polynucleotide phosphorylase-like protein, polyribonucleotide nucleotidyltransferase 1 | |
Mouse | |
86 kDa | |
100 μL | |
DNA replication Transcription Translation and Splicing | |
87178 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction