Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

pp9099 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant pp9099.

Manufacturer:  Abnova Corporation H00080301A01

Catalog No. 89-107-371


Add to cart

Description

Description

Sequence: GDLLGEGPRHPLQPRERLYRAQLEVKVASEQTEKLLNKVLGSEPAPVSAETLLSQAVEQLRQATQVLQEMRDLGELSQEAPGLREKRKELVTLYRRSAP
Specifications

Specifications

pp9099
Unconjugated
50% glycerol
NM_025201
DKFZp761K2312/FLJ38884/PLEKHQ1/PP1628/pp9099
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant pp9099.
PLEKHO2
Antisera
PLEKHO2
pp9099 (NP_079477, 392 a.a. ∽ 490 a.a) partial recombinant protein with GST tag.
RUO
Primary
80301
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only