Learn More
PPP2CA/PPP2CB, Mouse, Clone: 20G6, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of PPP2CA/PPP2CB.
Manufacturer:  Abnova Corporation MAB2678
Catalog No. 89-135-212
Add to cart
Description
Description
Specifications
Specifications
PPP2CA/PPP2CB
20G6
Mouse monoclonal antibody raised against synthetic peptide of PPP2CA/PPP2CB.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
Western Blot (1:2000) Immunoprecipitation (3 ul). The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to human PPP2CA/PPP2CB.
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal