Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP5C, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length recombinant PPP5C.

Manufacturer:  Abnova Corporation H00005536A01

Catalog No. 89-105-187


Add to cart

Description

Description

Sequence: MAMAEGERTECAEPPRDEPPADGALKRAEELKTQANDYFKAKDYENAIKFYSQAIELNPSNAIYYGNRSLAYLRTECYGYALGDATRAIELDKKYIKGYYRRAASNMALGKFRAALRDYETVVKVKPHDKDAKMKYQECNKIVKQKAFERAIAGDEHKRSVVDSLDIESMTIEDEYSGPKLEDGKVTISFMKELMQWYKDQKKLHRKCAYQILVQVKEVLSKLSTLVETTLKETEKITVCGDTHGQFYDLLNIFELNGLPSETNPYIFNGDFVDRGSFSVEVILTLFGFKLLYPDHFHLLRGNHETDNMNQIYGFEGEVKAKYTAQMYELFSEVFEWLPLAQCINGKVLIMHGGLFSEDGVTLDDIRKIERNRQPPDSGPMCDLLWSDPQPQNGRSISKRGVSCQFGPDVTKAFLEENNLDYIIRSHEVKAEGYEVAHGGRCVTVFSAPNYCDQMGNKASYIHLQGSDLRPQFHQFTAVPHPNVKPMAYANTLLQLGMM
Specifications

Specifications

PPP5C
Unconjugated
50% glycerol
BC001970
FLJ36922/PP5/PPP5
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
ELISA
Mouse polyclonal antibody raised against a full-length recombinant PPP5C.
PPP5C
Antisera
PPP5C
PPP5C (AAH01970, 1 a.a. ∽ 499 a.a) full-length recombinant protein with GST tag.
RUO
Primary
5536
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only