Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRDX1, Mouse, Clone: 13E7, Abnova™

Mouse monoclonal antibody raised against full-length recombinant PRDX1.

Manufacturer:  Abnova Corporation MAB0714

Catalog No. 89-108-070


Add to cart

Description

Description

This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8(+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. Three transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq

Specifications

Specifications

PRDX1
13E7
Mouse monoclonal antibody raised against full length recombinant PRDX1.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
PRDX1
Recombinant protein corresponding to full length human PRDX1.
Ammonium sulfate precipitation
RUO
Primary
5052
ELISA, Immunoprecipitation
Unconjugated
Immunoprecipitation (1-2 ul) The optimal working dilution should be determined by the end user.
PRDX1
MSP23/NKEFA/PAG/PAGA/PAGB/PRX1/PRXI/TDPX2
Mouse
IgG1, kappa
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit