Learn More
Prox1 Mouse anti-Human, Mouse, Rat, Chicken, DyLight 488, Clone: 5G10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP130045G
Catalog No. NBP130045G
Add to cart
Description
Description
Prox1 Monoclonal antibody specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
Prox1
5G10
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q92786
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1
Mouse
IgG1
0.1 ml
Vision
Primary
5629
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
PROX1
The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Chicken, Human, Mouse, Rat