Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVRIG, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PVRIG protein.

Manufacturer:  Abnova Corporation H00079037B01P

Catalog No. 89-107-314


Add to cart

Description

Description

Sequence: MRTEAQVPALQPPEPGLEGAMGHRTLVLPWVLLTLCVTAGTPEVWVQVRMEATELSSFTIRCGFLGSGSISLVTVSWGGPDGAGGTTLAVLHPERGIRQWAPARQARWETQSSISLILEGSGASSPCANTTFCCKFASFPEGSWEACGSLPPSSDPGLSAPPTPAPILRADLAGILGVSGVLLFGCVYLLHLLRRHKHRPAPRLQPSRTSPQAPRARAWAPSQASQAALHVPYATINTSCRPATLDTAHPHGGPSWWASLPTHAAHRPQGPAAWASTPIPARGSFVSVENGLYAQAGERPPHTGPGLTLFPDPRGPRAMEGPLGVR
Specifications

Specifications

PVRIG
Polyclonal
Mouse polyclonal antibody raised against a full-length human PVRIG protein.
PVRIG
Purified
PVRIG
PVRIG (AAH01129.1,1 a.a. ∽ 326 a.a) full-length human protein.
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
BC001129.2
C7orf15/MGC104322/MGC138295/MGC138297/MGC2463
Mouse
IgG
50 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
79037
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only