Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Rad51, Clone: Rad51 3C10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP1192560.2MG

Catalog No. NBP11925602


Add to cart

Description

Description

Rad51 Monoclonal antibody specifically detects Rad51 in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Rad51
3C10
Western Blot 1 ug/ml
Q06609
BRCC5, HRAD51, HsRAD51, HsT16930, RAD51 (S. cerevisiae) homolog (E coli RecA homolog), RAD51 homolog (RecA homolog, E. coli) (S. cerevisiae), RAD51 homolog A, RAD51ABRCA1/BRCA2-containing complex, subunit 5, RecA, E. coli, homolog of, RECADNA repair protein RAD51 homolog 1, RecA-like protein, recombination protein A
Mouse
IgG1
0.2 mg
Cancer, DNA Repair
Primary
5888
Western Blot
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Purified
RAD51
Recombinant human RAD51 protein
Protein G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only