Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD32 Mouse anti-Rat, Brilliant Violet 650, Clone: D34-485, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 740520

Catalog No. BDB740520


Add to cart

Description

Description

The D34-485 monoclonal antibody specifically recognizes CD32, the Fc γII receptor. Rat CD32 is expressed on B lymphocytes, myeloid cells, and some lymphocytes in the thymic medulla. D34-485 antibody blocks binding of aggregated immunoglobulins to the Fc γII receptors in vitro.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD32
D34-485
Brilliant Violet 650
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Fcgr2b; FcRII; Fc-gamma RII; FcγRII; FcγII receptor
Affinity Purified
Recombinant Rat CD32 Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only