Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8a Mouse anti-Rat, BUV395, Clone: OX-8, BD Optibuild™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 740257

Numéro de catalogue. BDB740257


ajouter au panier

Description

Description

The OX-8 monoclonal antibody specifically binds to the hinge-like membrane-proximal domain of the 32 kDa α chain of the CD8 differentiation antigen. A truncated CD8 α isoform has not been detected in the rat. The CD8 α and β chains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and a subpopulation of mature T lymphocytes (i.e., MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). Intestinal intrapithelial lymphocytes, many CD8+ T cells of athymic rats, many activated CD4+ T cells, and most NK cells express CD8a without CD8b. It has been suggested that the expression of the CD8a/CD8b heterodimer is restricted to thymus-derived T lymphocytes. OX-8 antibody does not react with resting CD4+ T helper cells. CD8 is an antigen coreceptor on the T-cell surface which interacts with MHC class I molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase Ick. Macrophages have also been reported to express CD8 α and β chains, which are involved in signal transduction. Soluble OX-8 mAb partially blocks in vitro MLR and CTL activity.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY
Spécifications

Spécifications

CD8a
Monoclonal
0.2mg/mL
Cd8a; CD8α; CD8 alpha; OX-8 membrane antigen
Mouse
Affinity Purified
RUO
Rat
IgG1 κ
Flow Cytometry
OX-8
Brilliant Ultraviolet 395
Aqueous buffered solution containing ≤0.09% sodium azide.
High-molecular-weight rat thymocyte glycoproteins
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
missing translation for 'recommendations'

missing translation for 'recommendations'

FDS
Documents

Documents

Product Certifications
Offres spéciales

Offres spéciales

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'

For Research Use Only