Learn More
Mouse anti-Reduced Folate Carrier/SLC19A1, APC, Clone: 890513, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB8450A025
Catalog No. FAB8450A025
Add to cart
This item is not returnable. View return policy
Description
Description
Reduced Folate Carrier/SLC19A1 Monoclonal antibody specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Reduced Folate Carrier/SLC19A1
890513
Flow Cytometry 10 uL/10^6 cells
P41440
CHMD, FLOT1, folate transporter 1, FOLT, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Stains human Reduced Folate Carrier/SLC19A1 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified, Supernatant
SLC19A1
NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1 Met1-Gln591 Accession # P41440
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6573
Human