Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RIPK1, Mouse, Clone: 7H10, Abnova™

Mouse monoclonal antibody raised against partial recombinant RIPK1.

Manufacturer:  Abnova Corporation MAB0836

Catalog No. 89-108-180


Add to cart

Description

Description

Specifications

Specifications

RIPK1
7H10
Mouse monoclonal antibody raised against partial recombinant RIPK1.
In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Liquid
RIPK1
Recombinant His/ABD fusion protein corresponding to amino acids 385-671 of human RIPK1.
Ammonium sulfate precipitation
RUO
Primary
8737
Immunoprecipitation, Western Blot
Unconjugated
Western Blot (1:500-1:2000) Immunoprecipitation (2 ul) The optimal working dilution should be determined by the end user.
RIPK1
FLJ39204/RIP/RIP1
Mouse
IgG2b, kappa
100 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit